Assay Details
| Gene Symbol: ddah1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127052.1 | NP_001120524.1 | 4 - 5 | 677 | 61 | |
| XM_018092936.1 | XP_017948425.1 | 4 - 5 | 838 | 61 | ||
| GenBank mRNA | BC124565.1 | - | 4 - 5 | 677 | 61 | |
More Information
Set Membership: |
|

ddah1
Genomic Map