Assay Details
| Gene Symbol: has2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127085.1 | NP_001120557.1 | 2 - 3 | 1153 | 82 | |
| GenBank mRNA | BC161505.1 | - | 2 - 3 | 1153 | 82 | |
More Information
Set Membership: |
|

has2
Genomic Map