Assay Details
| Gene Symbol: eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127089.2 | NP_001120561.2 | 10 - 11 | 1514 | 91 | |
| GenBank mRNA | CR587924.1 | - | 1 - 2 | 131 | 91 | |
| EL851385.1 | - | 4 - 5 | 557 | 91 | ||
More Information
Set Membership: |
|

eif2a
Genomic Map