Assay Details
| Gene Symbol: eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127089.2 | NP_001120561.2 | 11 - 12 | 1634 | 87 | |
| GenBank mRNA | CR587924.1 | - | 2 - 3 | 251 | 87 | |
| EL851385.1 | - | 3 - 4 | 437 | 87 | ||
More Information
Set Membership: |
|

eif2a
Genomic Map