Assay Details
| Gene Symbol: eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127089.2 | NP_001120561.2 | 13 - 14 | 1816 | 98 | |
| GenBank mRNA | CR587924.1 | - | 4 - 5 | 433 | 98 | |
| EL851385.1 | - | 1 - 2 | 255 | 98 | ||
More Information
Set Membership: |
|

eif2a
Genomic Map