Assay Details
| Gene Symbol: eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127089.2 | NP_001120561.2 | 2 - 3 | 235 | 74 | |
| GenBank mRNA | CX455947.2 | - | 2 - 3 | 236 | 74 | |
| DT404123.1 | - | 2 - 3 | 189 | 74 | ||
More Information
Set Membership: |
|

eif2a
Genomic Map