Assay Details
| Gene Symbol: eif2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127089.2 | NP_001120561.2 | 9 - 10 | 955 | 67 | |
| GenBank mRNA | CX343974.2 | - | 2 - 3 | 225 | 67 | |
More Information
Set Membership: |
|

eif2a
Genomic Map