Assay Details
| Gene Symbol: kdelc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127103.1 | NP_001120575.1 | 7 - 8 | 1446 | 77 | |
| GenBank mRNA | BC161552.1 | - | 7 - 8 | 1446 | 77 | |
| BC167365.1 | - | 7 - 8 | 1446 | 77 | ||
| BC168093.1 | - | 7 - 8 | 1446 | 77 | ||
More Information
Set Membership: |
|

kdelc2
Genomic Map