Assay Details
Gene Symbol: pkdcc.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127116.1 | NP_001120588.1 | 5 - 6 | 1282 | 84 | |
GenBank mRNA | BC161597.1 | - | 5 - 6 | 1282 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
