Assay Details
| Gene Symbol: mettl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127148.1 | NP_001120620.1 | 8 - 9 | 822 | 71 | |
| GenBank mRNA | BC161764.1 | - | 8 - 9 | 822 | 71 | |
| CU075325.1 | - | 8 - 9 | 826 | 71 | ||
More Information
Set Membership: |
|

mettl17
Genomic Map