Assay Details
| Gene Symbol: cipc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127149.1 | NP_001120621.1 | 3 - 4 | 396 | 85 | |
| XM_012968047.2 | XP_012823501.1 | 3 - 4 | 540 | 85 | ||
| XM_012968048.2 | XP_012823502.1 | 3 - 4 | 336 | 85 | ||
| GenBank mRNA | BC161765.1 | - | 3 - 4 | 396 | 85 | |
More Information
Set Membership: |
|

cipc
Genomic Map