Assay Details
| Gene Symbol: aldh3b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127150.1 | NP_001120622.1 | 2 - 3 | 284 | 80 | |
| GenBank mRNA | BC161766.1 | - | 2 - 3 | 284 | 80 | |
More Information
Set Membership: |
|

aldh3b2
Genomic Map