Assay Details
| Gene Symbol: gsr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127154.2 | NP_001120626.2 | 9 - 10 | 1304 | 75 | |
| GenBank mRNA | BC161777.1 | - | 9 - 10 | 1260 | 75 | |
More Information
Set Membership: |
|

gsr
Genomic Map