Assay Details
| Gene Symbol: pkmyt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127159.1 | NP_001120631.1 | 6 - 7 | 1481 | 71 | |
| XM_012970105.2 | XP_012825559.1 | 6 - 7 | 1566 | 71 | ||
| XM_012970106.2 | XP_012825560.1 | 6 - 7 | 1532 | 71 | ||
| XM_012970107.2 | XP_012825561.1 | 6 - 7 | 2343 | 71 | ||
| GenBank mRNA | BC161784.1 | - | 6 - 7 | 1481 | 71 | |
More Information
Set Membership: |
|

pkmyt1
Genomic Map