Assay Details
| Gene Symbol: parp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127163.2 | NP_001120635.1 | 6 - 7 | 920 | 74 | |
| XM_012960640.2 | XP_012816094.1 | 6 - 7 | 1022 | 74 | ||
| GenBank mRNA | BC161791.1 | - | 6 - 7 | 922 | 74 | |
More Information
Set Membership: |
|

parp3
Genomic Map