Assay Details
| Gene Symbol: kcnh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127437.1 | NP_001120909.1 | 8 - 9 | 1685 | 76 | |
| XM_012968051.2 | XP_012823505.1 | 9 - 10 | 2202 | 76 | ||
| GenBank mRNA | BC166159.1 | - | 8 - 9 | 1685 | 76 | |
More Information
Set Membership: |
|

kcnh5
Genomic Map