Assay Details
| Gene Symbol: retreg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127953.1 | NP_001121425.1 | 2 - 3 | 515 | 90 | |
| GenBank mRNA | BC166162.1 | - | 2 - 3 | 515 | 90 | |
More Information
Set Membership: |
|

retreg2
Genomic Map