Assay Details
| Gene Symbol: ropn1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127968.1 | NP_001121440.1 | 2 - 3 | 267 | 83 | |
| GenBank mRNA | BC166191.1 | - | 2 - 3 | 267 | 83 | |
More Information
Set Membership: |
|

ropn1l
Genomic Map