Assay Details
| Gene Symbol: plpp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128006.2 | NP_001121478.1 | 2 - 3 | 173 | 96 | |
| GenBank mRNA | AL888583.2 | - | 2 - 3 | 123 | 96 | |
| BC166252.1 | - | 2 - 3 | 126 | 96 | ||
| BC168452.1 | - | 1 - 2 | 60 | 96 | ||
| CU024953.1 | - | 1 - 2 | 102 | 96 | ||
| CX917894.1 | - | 2 - 3 | 164 | 96 | ||
More Information
Set Membership: |
|

plpp5
Genomic Map