Assay Details
| Gene Symbol: nptx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128007.1 | NP_001121479.1 | 2 - 3 | 725 | 69 | |
| GenBank mRNA | BC166253.1 | - | 2 - 3 | 725 | 69 | |
More Information
Set Membership: |
|

nptx1
Genomic Map