Assay Details
| Gene Symbol: aste1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128015.1 | NP_001121487.1 | 4 - 5 | 1858 | 66 | |
| GenBank mRNA | BC075288.1 | - | 1878 | 66 | ||
| BC166275.1 | - | 4 - 5 | 1858 | 66 | ||
| CR926284.2 | - | 4 - 5 | 1892 | 66 | ||
More Information
Set Membership: |
|

aste1
Genomic Map