Assay Details
Gene Symbol: tm6sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128019.1 | NP_001121491.1 | 3 - 4 | 348 | 70 | |
XM_018091177.1 | XP_017946666.1 | 2 - 3 | 187 | 70 | ||
GenBank mRNA | BC166284.1 | - | 3 - 4 | 348 | 70 | |
CU075900.1 | - | 3 - 4 | 321 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
