Assay Details
| Gene Symbol: nuak2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128027.1 | NP_001121499.1 | 1 - 2 | 194 | 85 | |
| GenBank mRNA | BC166319.1 | - | 1 - 2 | 194 | 85 | |
More Information
Set Membership: |
|

nuak2
Genomic Map