Assay Details
| Gene Symbol: fam171a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128032.2 | NP_001121504.2 | 4 - 5 | 623 | 57 | |
| XM_012971549.2 | XP_012827003.1 | 4 - 5 | 728 | 57 | ||
| GenBank mRNA | BC166337.1 | - | 4 - 5 | 623 | 57 | |
More Information
Set Membership: |
|

fam171a2
Genomic Map