Assay Details
| Gene Symbol: raver1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128048.1 | NP_001121520.1 | 4 - 5 | 982 | 78 | |
| GenBank mRNA | BC166376.1 | - | 4 - 5 | 982 | 78 | |
| CT030621.1 | - | 985 | 78 | |||
More Information
Set Membership: |
|

raver1
Genomic Map