Assay Details
| Gene Symbol: gpr153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128052.1 | NP_001121524.1 | 2 - 3 | 887 | 96 | |
| XM_012966140.2 | XP_012821594.1 | 3 - 4 | 1065 | 96 | ||
| GenBank mRNA | BC166386.1 | - | 2 - 3 | 887 | 96 | |
More Information
Set Membership: |
|

gpr153
Genomic Map