Assay Details
| Gene Symbol: hhla2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128644.1 | NP_001122116.1 | 4 - 5 | 887 | 78 | |
| GenBank mRNA | BC096014.1 | - | 3 - 4 | 679 | 78 | |
| BC167398.1 | - | 4 - 5 | 887 | 78 | ||
More Information
Set Membership: |
|

hhla2
Genomic Map