Assay Details
| Gene Symbol: sftpc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128660.2 | NP_001122132.2 | 2 - 3 | 197 | 67 | |
| GenBank mRNA | BC089701.1 | - | 2 - 3 | 193 | 67 | |
| BC167703.1 | - | 2 - 3 | 197 | 67 | ||
More Information
Set Membership: |
|

sftpc
Genomic Map