Assay Details
| Gene Symbol: pbx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128661.1 | NP_001122133.1 | 1 - 2 | 659 | 91 | |
| XM_012967841.2 | XP_012823295.1 | 1 - 2 | 719 | 91 | ||
| XM_018096098.1 | XP_017951587.1 | 1 - 2 | 719 | 91 | ||
| GenBank mRNA | BC158305.1 | - | 1 - 2 | 553 | 91 | |
| BC167699.1 | - | 1 - 2 | 659 | 91 | ||
More Information
Set Membership: |
|

pbx2
Genomic Map