Assay Details
| Gene Symbol: LOC100170420 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130198.1 | NP_001123670.1 | 2 - 3 | 163 | 95 | |
| GenBank mRNA | BC166957.1 | - | 2 - 3 | 163 | 95 | |
| CT025446.2 | - | 2 - 3 | 129 | 95 | ||
More Information
Set Membership: |
|

LOC100170420
Genomic Map