Assay Details
| Gene Symbol: ak3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130221.1 | NP_001123693.1 | 1 - 2 | 203 | 74 | |
| XM_012956966.2 | XP_012812420.1 | 1 - 2 | 468 | 74 | ||
| GenBank mRNA | BC167275.1 | - | 1 - 2 | 203 | 74 | |
More Information
Set Membership: |
|

ak3
Genomic Map