Assay Details
| Gene Symbol: epm2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130223.1 | NP_001123695.1 | 4 - 5 | 785 | 71 | |
| XM_012962766.2 | XP_012818220.1 | 4 - 5 | 768 | 71 | ||
| GenBank mRNA | BC167294.1 | - | 4 - 5 | 785 | 71 | |
| BC167295.1 | - | 4 - 5 | 785 | 71 | ||
More Information
Set Membership: |
|

epm2a
Genomic Map