Assay Details
| Gene Symbol: lto1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130228.1 | NP_001123700.1 | 4 - 5 | 365 | 100 | |
| GenBank mRNA | BC167301.1 | - | 4 - 5 | 365 | 100 | |
More Information
Set Membership: |
|

lto1
Genomic Map