Assay Details
| Gene Symbol: cry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130234.1 | NP_001123706.1 | 4 - 5 | 741 | 87 | |
| XM_012956989.2 | XP_012812443.1 | 4 - 5 | 888 | 87 | ||
| GenBank mRNA | AB534559.1 | - | 3 - 4 | 688 | 87 | |
| BC167313.1 | - | 4 - 5 | 741 | 87 | ||
More Information
Set Membership: |
|

cry4
Genomic Map