Assay Details
| Gene Symbol: cry4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130234.1 | NP_001123706.1 | 7 - 8 | 1197 | 100 | |
| XM_012956989.2 | XP_012812443.1 | 7 - 8 | 1344 | 100 | ||
| GenBank mRNA | AB534559.1 | - | 6 - 7 | 1144 | 100 | |
| BC167313.1 | - | 7 - 8 | 1197 | 100 | ||
More Information
Set Membership: |
|

cry4
Genomic Map