Assay Details
| Gene Symbol: spart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130255.1 | NP_001123727.1 | 2 - 3 | 1032 | 84 | |
| XM_012956537.2 | XP_012811991.1 | 3 - 4 | 1200 | 84 | ||
| GenBank mRNA | BC167368.1 | - | 2 - 3 | 1032 | 84 | |
More Information
Set Membership: |
|

spart
Genomic Map