Assay Details
| Gene Symbol: spart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130255.1 | NP_001123727.1 | 5 - 6 | 1506 | 67 | |
| XM_012956537.2 | XP_012811991.1 | 6 - 7 | 1674 | 67 | ||
| GenBank mRNA | BC167368.1 | - | 5 - 6 | 1506 | 67 | |
More Information
Set Membership: |
|

spart
Genomic Map