Assay Details
| Gene Symbol: spart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130255.1 | NP_001123727.1 | 7 - 8 | 1648 | 95 | |
| GenBank mRNA | BC167368.1 | - | 7 - 8 | 1648 | 95 | |
More Information
Set Membership: |
|

spart
Genomic Map