Assay Details
| Gene Symbol: panx1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130256.1 | NP_001123728.1 | 3 - 4 | 649 | 66 | |
| GenBank mRNA | BC167372.1 | - | 3 - 4 | 649 | 66 | |
More Information
Set Membership: |
|

panx1
Genomic Map