Assay Details
| Gene Symbol: lrrc39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130264.1 | NP_001123736.1 | 2 - 3 | 499 | 69 | |
| GenBank mRNA | BC167387.1 | - | 2 - 3 | 499 | 69 | |
More Information
Set Membership: |
|

lrrc39
Genomic Map