Assay Details
| Gene Symbol: ripor2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130276.1 | NP_001123748.1 | 12 - 13 | 1304 | 65 | |
| GenBank mRNA | BC167462.1 | - | 12 - 13 | 1304 | 65 | |
More Information
Set Membership: |
|

ripor2
Genomic Map