Assay Details
| Gene Symbol: LOC101733051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004918304.3 | XP_004918361.3 | 4 - 5 | 418 | 69 | |
| Gene Symbol: tsc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012970165.2 | XP_012825619.1 | 18 - 19 | 2052 | 69 | |
| XM_012970166.2 | XP_012825620.1 | 18 - 19 | 2052 | 69 | ||
| GenBank mRNA | BC167586.1 | - | 18 - 19 | 2096 | 69 | |
| CT030619.1 | - | 18 - 19 | 2005 | 69 | ||
More Information
Set Membership: |
|

LOC101733051
Genomic Map