Assay Details
| Gene Symbol: rpl27a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130329.1 | NP_001123801.1 | 2 - 3 | 414 | 62 | |
| XM_012966164.1 | XP_012821618.1 | 2 - 3 | 249 | 62 | ||
| GenBank mRNA | BC167596.1 | - | 2 - 3 | 414 | 62 | |
| BC168012.1 | - | 2 - 3 | 414 | 62 | ||
More Information
Set Membership: |
|

rpl27a
Genomic Map