Assay Details
| Gene Symbol: gpat4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130350.1 | NP_001123822.1 | 10 - 11 | 1757 | 75 | |
| XM_012958656.2 | XP_012814110.1 | 11 - 12 | 1915 | 75 | ||
| XM_012958657.2 | XP_012814111.1 | 11 - 12 | 1942 | 75 | ||
| GenBank mRNA | BC167630.1 | - | 10 - 11 | 1757 | 75 | |
More Information
Set Membership: |
|

gpat4
Genomic Map