Assay Details
| Gene Symbol: cpz | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130361.1 | NP_001123833.1 | 7 - 8 | 1120 | 73 | |
| XM_012957083.2 | XP_012812537.1 | 9 - 10 | 1634 | 73 | ||
| XM_012957091.2 | XP_012812545.1 | 8 - 9 | 1604 | 73 | ||
| GenBank mRNA | BC167656.1 | - | 7 - 8 | 1120 | 73 | |
More Information
Set Membership: |
|

cpz
Genomic Map