Assay Details
| Gene Symbol: LOC100170623 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018091853.1 | XP_017947342.1 | 5 - 6 | 591 | 74 | |
| GenBank mRNA | BC167735.1 | - | 5 - 6 | 578 | 74 | |
More Information
Set Membership: |
|

LOC100170623
Genomic Map