Assay Details
| Gene Symbol: ndufa11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130386.1 | NP_001123858.1 | 3 - 4 | 327 | 98 | |
| XM_012957126.2 | XP_012812580.1 | 3 - 4 | 390 | 98 | ||
| GenBank mRNA | BC167739.1 | - | 3 - 4 | 327 | 98 | |
More Information
Set Membership: |
|

ndufa11
Genomic Map