Assay Details
| Gene Symbol: psmc3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130530.1 | NP_001124002.1 | 7 - 8 | 658 | 69 | |
| XM_012966161.1 | XP_012821615.1 | 6 - 7 | 560 | 69 | ||
| GenBank mRNA | BC167145.1 | - | 7 - 8 | 658 | 69 | |
More Information
Set Membership: |
|

psmc3ip
Genomic Map