Assay Details
| Gene Symbol: gpr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131040.1 | NP_001124512.1 | 5 - 6 | 877 | 65 | |
| GenBank mRNA | BC168132.1 | - | 5 - 6 | 877 | 65 | |
| CR926383.2 | - | 4 - 5 | 874 | 65 | ||
More Information
Set Membership: |
|

gpr157
Genomic Map