Assay Details
| Gene Symbol: gpnmb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001131042.1 | NP_001124514.1 | 6 - 7 | 1227 | 72 | |
| GenBank mRNA | BC168052.1 | - | 6 - 7 | 1227 | 72 | |
| CU075468.1 | - | 6 - 7 | 1049 | 72 | ||
More Information
Set Membership: |
|

gpnmb
Genomic Map